Learn More
Volu Sol Dehydrant 100% -(128 oz / 3.78 L), Case of 4
Supplier: Volu Sol VDH128C
Dehydrant 100, dehydrants are a blend of methyl, ethyl, and isopropyl alcohols, available in four different concentrations. Specifically formulated for use in processing and staining as dye or stain solvents in place of ethyl alcohol. Miscible with all clearing reagents and can be used in histology and cytology procedures