Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrated Alcohol, 200 Proof, BiotechGrade, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18602021 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-021 500 mL Amber Glass Bottle
18-602-022 4 L Amber Glass Bottle
18-608-003 200 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-021 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1424500ML
Add to Cart
Add to Cart

E1424, 64-17-5, C2H5OH

Spectrum™ Chemical Dehydrated Alcohol, 200 Proof, BiotechGrade The GR is an EMD trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
Water 0.2%
UN Number UN1170
Concentration 100%
Boiling Point 78°C to 79°C
Percent Purity 99.5%
Grade Biotech
Chemical Name or Material Alcohol
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.