Learn More
Dehydrated Alcohol, 200 Proof, EP, BP, USP, 99.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Spectrum™ Chemical Dehydrated Alcohol, 200 Proof, USP, EP, BP is used as an antiseptic to disinfect skin prior to injections, an active ingredient for pain relief and as a solvent. All Spectrum Chemical USP, EP, BP dehydrated alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| Specific Gravity | 0.7962 |
| UN Number | UN1170 |
| Concentration | 100% |
| Boiling Point | 78°C to 79°C |
| Percent Purity | 99.5% |
| Grade | Multi-Compendial |
| Chemical Name or Material | Alcohol |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.