Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrated Alcohol, 200 Proof, Undenatured, USP, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18602024 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-024 500 mL Amber Glass Bottle
18-602-023 100 mL Amber Glass Bottle
18-602-026 1 L Amber Glass Bottle
18-602-025 6 x 500 mL Amber Glass Bottle
18-602-027 4 L Amber Glass Bottle
18-602-028 6 x 1 L Amber Glass Bottle
18-602-029 4 x 4 L Amber Glass Bottle
18-602-030 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-602-024 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ET107500MLGL
Only null left
Add to Cart
Add to Cart

ET107, 64-17-5, C2H5OH

Spectrum™ Chemical Dehydrated Alcohol, 200 Proof, Undenatured, USP is used as an antiseptic to disinfect skin prior to injections, an active ingredient for pain relief and as a solvent. Spectrum manufacturers Dehydrated Alcohol, 200 Proof, Undenatured, USP and packages it in sizes that various sizes from convenient laboratory 100 mL bottles to 200 L production drums. Like all Spectrum Chemical USP products, dehydrated alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
Specific Gravity 0.7962
UN Number UN1170
Concentration 100%
Boiling Point 78°C to 79°C
Percent Purity 99.5%
Grade USP
Chemical Name or Material Alcohol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.