Learn More
Enzo Life Sciences Dehydrin (plant) polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIPLA100D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of dehydrin. The sequence is completely conserved in barley, rice, maize and wheat. UniProt: A2ZDX9 (rice dehydrin family member Rab21). Source: Purified from rabbit serum. Species Reactivity: Plant. Applications: WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-345