Learn More
Enzo Life Sciences Dehydrin (plant) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIPLA100F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of dehydrin. The sequence is completely conserved in barley, rice, maize and wheat. Source: Purified from rabbit serum. Species reactivity: Plant. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7388