Learn More
Cayman Chemical Dehydro Amlodipine (fumarate), Purity: ≥98%, CAS: 2138811-33-1, 10 mg
SDP
Catalog No. 502932678
Encompass_Preferred
A potential impurity found in commercial preparations of amlodipine; a degradation product formed during storage or when amlodipine is subjected to oxidative or acidic conditions or undergoes photo or electrochemical degradation
Catalog No. 50-293-2678 Supplier Cayman Chemical Supplier No. 2662210 MG
A potential impurity found in commercial preparations of amlodipine; a degradation product formed during storage or when amlodipine is subjected to oxidative or acidic conditions or undergoes photo or electrochemical degradation