Learn More
Cayman Chemical Dehydro NIfedIpIn 10mg
SDP
Catalog No. 502987211
Encompass_Preferred
An active metabolite of nifedipine; formed when nifedipine is metabolized by CYP3A4 and CYP3A5; inhibits glucose uptake in PC-12 cells (IC50 = 130 μM)
Catalog No. 50-298-7211 Supplier Cayman Chemical Supplier No. 2322910 MG
An active metabolite of nifedipine; formed when nifedipine is metabolized by CYP3A4 and CYP3A5; inhibits glucose uptake in PC-12 cells (IC50 = 130 μM)