Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dehydro olmesartan | 172875-98-8 | MFCD18379313 | 99.4% | 428.49 g/mol | C24H24N6O2 | 5 MG
SDP

Catalog No. 5000262978
Encompass_Preferred

Dehydro olmesartan is a chemical derivative of olmesartan that acts as an angiotensin II type 1 receptor (AT1R) antagonist. Supplied as a powder for laboratory research, it is intended for in vitro and preclinical studies related to blood pressure regulation and angiotensin receptor biology. For research use only; not validated for clinical applications.

  • High reported purity (99.43%).
  • Molecular formula C24H24N6O2; molecular weight 428.49 g/mol.
  • Supplied as a powder; storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Biological target: angiotensin receptor (AT1R); pathway: GPCR/G protein.
  • Available in small research quantities (e.g., 5 mg).

Catalog No. 50-002-62978 Supplier Medchemexpress LLC Supplier No. HY1312775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dehydro olmesartan is a chemical derivative of olmesartan that acts as an angiotensin II type 1 receptor (AT1R) antagonist. Supplied as a powder for laboratory research, it is intended for in vitro and preclinical studies related to blood pressure regulation and angiotensin receptor biology. For research use only; not validated for clinical applications.

  • High reported purity (99.43%).
  • Molecular formula C24H24N6O2; molecular weight 428.49 g/mol.
  • Supplied as a powder; storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Biological target: angiotensin receptor (AT1R); pathway: GPCR/G protein.
  • Available in small research quantities (e.g., 5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.