Learn More
Medchemexpress LLC HY-103061 5mg Medchemexpress, Dehydro-ZINC39395747 CAS:433248-87-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1030615MG
Encompass_Preferred
Medchemexpress, HY-103061 5mg Dehydro-ZINC39395747 CAS:433248-87-4 Dehydro-ZINC39395747 is a derivative of ZINC39395747. ZINC39395747 is a potent cytochrome b5 reductase 3 (CYB5R3) inhibitor with an IC50 of 9.14 μM and a Kd of 1.11 μM. ZINC39395747 can increase NO bioavailability in vascular cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9520