Learn More
Selleck Chemical LLC Dehydroaripiprazole E7427-5MG
SDP
Catalog No. 5000041822
Encompass_Preferred
Dehydroaripiprazole is the active metabolite of aripiprazole primarily metabolized by CYP3A4 and CYP2D6 It inhibits glutamate release from rat prefrontocortical nerve terminals by activating dopamine D2 and 5-HT1A receptors reducing nerve terminal excitability and VDCC activation through a PKA-mediated signaling cascade potentially contributing to its neuroprotective effects in excitotoxic injury
Catalog No. 50-000-41822 Supplier Selleck Chemical LLC Supplier No. E74275MG
Dehydroaripiprazole is the active metabolite of aripiprazole primarily metabolized by CYP3A4 and CYP2D6 It inhibits glutamate release from rat prefrontocortical nerve terminals by activating dopamine D2 and 5-HT1A receptors reducing nerve terminal excitability and VDCC activation through a PKA-mediated signaling cascade potentially contributing to its neuroprotective effects in excitotoxic injury