Learn More
Cayman Chemical DehydroascorbIc AcId 100mg
SDP
Catalog No. 502938175
Encompass_Preferred
An oxidized form of ascorbic acid; inhibits hydrogen peroxide-induced cell death in murine astrocytes by approximately 74 and 83% at 0.5 and 1 mM, respectively; increases GPX and glutathione reductase activity and inhibits ROS production in hydrogen peroxide-treated astrocytes at 1 mM; increases cerebral blood flow and reduces infarct volume and mortality in a mouse model of cerebral ischemia-reperfusion injury induced by transient MCAO when administered prior to ischemia at 40 and 250 mg/kg; reduces infarct volume in a permanent MCAO mouse model when administered pre- or post-ischemia at 250 and 500 mg/kg, respectively
Catalog No. 50-293-8175 Supplier Cayman Chemical Supplier No. 29089100 MG
An oxidized form of ascorbic acid; inhibits hydrogen peroxide-induced cell death in murine astrocytes by approximately 74 and 83% at 0.5 and 1 mM, respectively; increases GPX and glutathione reductase activity and inhibits ROS production in hydrogen peroxide-treated astrocytes at 1 mM; increases cerebral blood flow and reduces infarct volume and mortality in a mouse model of cerebral ischemia-reperfusion injury induced by transient MCAO when administered prior to ischemia at 40 and 250 mg/kg; reduces infarct volume in a permanent MCAO mouse model when administered pre- or post-ischemia at 250 and 500 mg/kg, respectively