Learn More
Medchemexpress LLC Dehydroascorbic acid | 490-83-5 | 95.0% | 174.11 | 1 ML
SDP
Catalog No. 5000343952
Encompass_Preferred
Dehydroascorbic acid is an oxidized form of vitamin C that can cross the blood-brain barrier (BBB). It clears cytotoxic reactive oxygen species (ROS) produced after ischemic stroke by converting to ascorbic acid (AA), thereby reducing neuronal and glial cell damage and stabilizing cerebral microvascular NO signaling to maintain perfusion in the ischemic area. It can be used in research on ischemic stroke.
  • Crosses the blood-brain barrier
  • Clears cytotoxic reactive oxygen species (ROS)
  • Reduces neuronal and glial cell damage
  • Stabilizes cerebral microvascular NO signaling
  • Maintains perfusion in the ischemic area
  • Used in research on ischemic stroke
Catalog No. 50-003-43952 Supplier Medchemexpress LLC Supplier No. HY110281100MG
Dehydroascorbic acid is an oxidized form of vitamin C that can cross the blood-brain barrier (BBB). It clears cytotoxic reactive oxygen species (ROS) produced after ischemic stroke by converting to ascorbic acid (AA), thereby reducing neuronal and glial cell damage and stabilizing cerebral microvascular NO signaling to maintain perfusion in the ischemic area. It can be used in research on ischemic stroke.
  • Crosses the blood-brain barrier
  • Clears cytotoxic reactive oxygen species (ROS)
  • Reduces neuronal and glial cell damage
  • Stabilizes cerebral microvascular NO signaling
  • Maintains perfusion in the ischemic area
  • Used in research on ischemic stroke