Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrocholic Acid, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18602034 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-034 25 g Poly Bottle
18-602-035 100 g Poly Bottle
18-602-036 500 g Poly Pail
18-602-038 1 kg Poly Pail
18-602-037 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-034 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D100125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

D1001, 81-23-2, C24H34O5

Spectrum™ Chemical Dehydrocholic Acid, USP is used for the treatment of Biliary-tract disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 81-23-2
Molecular Formula C24H34O5
Molecular Weight (g/mol) 402.53
InChI Key OHXPGWPVLFPUSM-KLRNGDHRSA-N
IUPAC Name (4R)-4-[(1R,3aS,3bR,5aS,9aS,9bS,11aR)-9a,11a-dimethyl-4,7,11-trioxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
SMILES C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]1(C)CCC(=O)C[C@H]1CC3=O

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 1%
Melting Range 231-242°C
Packaging Poly Bottle
Quantity 25 g
Specific Rotation +29.0 to +32.5°
Percent Purity 98.5 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.