Learn More
Cayman Chemical Dehydrocannabifuran, Purity: ≥95%, CAS: 56154-59-7, 5 mg
SDP
Catalog No. 502944569
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis and marijuana smoke condensate; intended for research and forensic applications
Catalog No. 50-294-4569 Supplier Cayman Chemical Supplier No. 397825 MG
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis and marijuana smoke condensate; intended for research and forensic applications