Learn More
Medchemexpress LLC HY-N0674 5mg Medchemexpress, Dehydrocorydaline CAS:30045-16-0 Purity:>98%
SDP
Catalog No. 501933670
Encompass_Preferred
Medchemexpress, HY-N0674 5mg Dehydrocorydaline CAS:30045-16-0 Dehydrocorydaline (13-Methylpalmatine) is an alkaloid isolated from traditional Chinese herb Corydalis yanhusuo W.T. Wang. Dehydrocorydaline regulates protein expression of Bax , Bcl-2 ; activates caspase-7 , caspase-8 , and inactivates PARP . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3670 Supplier Medchemexpress LLC Supplier No. HYN06745MG
Medchemexpress, HY-N0674 5mg Dehydrocorydaline CAS:30045-16-0 Dehydrocorydaline (13-Methylpalmatine) is an alkaloid isolated from traditional Chinese herb Corydalis yanhusuo W.T. Wang. Dehydrocorydaline regulates protein expression of Bax , Bcl-2 ; activates caspase-7 , caspase-8 , and inactivates PARP . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.