Learn More
Medchemexpress LLC HY-N0674A 5mg Medchemexpress, Dehydrocorydaline (chloride) CAS:10605-03-5 Purity:>98%
SDP
Catalog No. 502031376
Encompass_Preferred
Medchemexpress, HY-N0674A 5mg Dehydrocorydaline (chloride) CAS:10605-03-5 Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid isolated from traditional Chinese herb Corydalis yanhusuo W.T. Wang. Dehydrocorydaline regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. Dehydrocorydaline chloride elevates p38 MAPK activation. Anti-inflammatory and anti-cancer activities.[2]. Dehydrocorydaline chloride shows strong anti-malarial effects (IC50 =38 nM), and low cytotoxicity (cell viability > 90%) using P. falciparum 3D7 strain[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1376 Supplier Medchemexpress LLC Supplier No. HYN0674A5MG
Medchemexpress, HY-N0674A 5mg Dehydrocorydaline (chloride) CAS:10605-03-5 Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid isolated from traditional Chinese herb Corydalis yanhusuo W.T. Wang. Dehydrocorydaline regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. Dehydrocorydaline chloride elevates p38 MAPK activation. Anti-inflammatory and anti-cancer activities.[2]. Dehydrocorydaline chloride shows strong anti-malarial effects (IC50 =38 nM), and low cytotoxicity (cell viability > 90%) using P. falciparum 3D7 strain[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.