Learn More
Medchemexpress LLC Dehydrocorydaline nitrate | 13005-09-9 | 99.9% | 428.44 g/mol | C22H24N2O7 | 10 MG
SDP
Catalog No. 5000289796
Encompass_Preferred
Dehydrocorydaline nitrate is a bioactive natural alkaloid used in research applications. It modulates apoptosis-related proteins and caspase activation, elevates p38 MAPK signaling, and has reported anti-inflammatory, anti-cancer, and potent antimalarial activity with low observed cytotoxicity.
  • Regulates apoptosis-related proteins and caspase activation.
  • Elevates p38 MAPK activation involved in stress response.
  • Reported anti-inflammatory and anti-cancer bioactivity.
  • Potent antimalarial activity with low observed cytotoxicity in tested strains.
  • High reported purity suitable for biochemical studies.
  • Stable solid form with defined storage recommendations for stability.
Catalog No. 50-002-89796 Supplier Medchemexpress LLC Supplier No. HYN423810MG
Dehydrocorydaline nitrate is a bioactive natural alkaloid used in research applications. It modulates apoptosis-related proteins and caspase activation, elevates p38 MAPK signaling, and has reported anti-inflammatory, anti-cancer, and potent antimalarial activity with low observed cytotoxicity.
  • Regulates apoptosis-related proteins and caspase activation.
  • Elevates p38 MAPK activation involved in stress response.
  • Reported anti-inflammatory and anti-cancer bioactivity.
  • Potent antimalarial activity with low observed cytotoxicity in tested strains.
  • High reported purity suitable for biochemical studies.
  • Stable solid form with defined storage recommendations for stability.