Learn More
Apexbio Technology LLC Dehydrocostus Lactone 477-43-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062825
Encompass_Preferred
Dehydrocostus lactone is a natural sesquiterpene lactone isolated from the plant Vladimiria souliei (Franch ) Ling Chemically this compound is characterized by an -methylene- -lactone ring structure which confers bioactivity through covalent modification of nucleophilic residues in target proteins Current research explores its potential biological roles in anti-inflammatory anti-tumor and immunomodulatory contexts In pharmacological studies Dehydrocostus lactone exhibits inhibitory activity toward NF- B signaling pathways thus affecting downstream cytokine and inflammatory mediator expression Additionally this sesquiterpene lactone has been investigated in vitro for its pro-apoptotic and anti-proliferative effects in various cancer cell lines Stored under sealed cool dry laboratory conditions it represents an active compound frequently utilized in biochemical assays and biomedical investigation
Catalog No. 50-000-62825 Supplier Apexbio Technology LLC Supplier No. N23345.1
Dehydrocostus lactone is a natural sesquiterpene lactone isolated from the plant Vladimiria souliei (Franch ) Ling Chemically this compound is characterized by an -methylene- -lactone ring structure which confers bioactivity through covalent modification of nucleophilic residues in target proteins Current research explores its potential biological roles in anti-inflammatory anti-tumor and immunomodulatory contexts In pharmacological studies Dehydrocostus lactone exhibits inhibitory activity toward NF- B signaling pathways thus affecting downstream cytokine and inflammatory mediator expression Additionally this sesquiterpene lactone has been investigated in vitro for its pro-apoptotic and anti-proliferative effects in various cancer cell lines Stored under sealed cool dry laboratory conditions it represents an active compound frequently utilized in biochemical assays and biomedical investigation