Learn More
TARGETMOL CHEMICALS INC DEHYDROCOSTUS LACTONE 25MG
SDP
Catalog No. 502577042
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dehydrocostus Lactone ((-)-Dehydrocostus lactone) shows anti-inflammatory anti-ulcer immunomodulatory and anti-tumor properties. purity: 99%
Catalog No. 50-257-7042 Supplier TARGETMOL CHEMICALS INC Supplier No. T283325MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dehydrocostus Lactone ((-)-Dehydrocostus lactone) shows anti-inflammatory anti-ulcer immunomodulatory and anti-tumor properties. purity: 99%