Learn More
Selleck Chemical LLC Dehydrocostus Lactone 25mg 477-43-0
SDP
Supplier:  Selleck Chemical LLC S361525MG
Encompass_Preferred
Dehydrocostus lactone (DHE), a natural sesquiterpene lactone, inhibits IKK? activity, I?B? phosphorylation and degradation, coactivators p300 recruitments and p50/p65 NF-?B nuclear translocation, and their DNA binding activity on COX-2 promoter. It has anti-inflammatory, anti-ulcer, immunomodulatory and anti-tumor properties. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8386