Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dehydrodiisoeugenol | 2680-81-1 | 100.0% | 326.39 g/mol | C20H22O4 | 10 MG
SDP

Catalog No. 5000296441
Encompass_Preferred

Dehydrodiisoeugenol is a natural phenylpropanoid compound used as a COX inhibitor and analytical research standard. It is supplied as a high-purity solid for biochemical, pharmacological, and analytical studies.

  • High purity 99.98%.
  • CAS number 2680-81-1.
  • Molecular formula C20H22O4; molecular weight 326.39 g/mol.
  • Available in small quantities suitable for analytical use (for example, 10 mg).
  • Includes datasheet, SDS, and certificate of analysis for quality verification.
  • Recommended storage: powder at -20°C for long-term; in solvent at -80°C for extended storage.

Catalog No. 50-002-96441 Supplier Medchemexpress LLC Supplier No. HYN058910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dehydrodiisoeugenol is a natural phenylpropanoid compound used as a COX inhibitor and analytical research standard. It is supplied as a high-purity solid for biochemical, pharmacological, and analytical studies.

  • High purity 99.98%.
  • CAS number 2680-81-1.
  • Molecular formula C20H22O4; molecular weight 326.39 g/mol.
  • Available in small quantities suitable for analytical use (for example, 10 mg).
  • Includes datasheet, SDS, and certificate of analysis for quality verification.
  • Recommended storage: powder at -20°C for long-term; in solvent at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.