Learn More
Medchemexpress LLC Dehydrodiisoeugenol | 2680-81-1 | 326.39 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN058920MG
Encompass_Preferred
Dehydrodiisoeugenol is an orally active anti-inflammatory and anti-tumor agent. It inhibits the proliferation of colorectal cancer cells and induces apoptosis, autophagy, endoplasmic reticulum stress, and cell cycle arrest. It also has anti-inflammatory effects by inhibiting the activation of NF-κB and the expression of COX-2. It can be used in research related to colorectal cancer, inflammatory diseases, and ulcerative colitis.
  • Orally active anti-inflammatory and anti-tumor agent.
  • Inhibits proliferation of colorectal cancer cells.
  • Induces apoptosis, autophagy, endoplasmic reticulum stress, and cell cycle arrest.
  • Exerts anti-inflammatory effects by inhibiting NF-κB activation and COX-2 expression.
  • Can be used in research for colorectal cancer, inflammatory diseases, and ulcerative colitis.
  • Purity: 99.98%.
Catalog No. 50-003-88498