Learn More
Medchemexpress LLC Dehydrodiisoeugenol | 2680-81-1 | 326.39 | 5 MG
SDP
Catalog No. 5000388450
Encompass_Preferred
Dehydrodiisoeugenol is an orally active compound demonstrating anti-inflammatory and anti-tumor properties. It is utilized in research focusing on colorectal cancer, inflammatory conditions, and ulcerative colitis.
  • Inhibits proliferation of colorectal cancer cells.
  • Induces apoptosis, autophagy, and cell cycle arrest.
  • Exerts anti-inflammatory effects by inhibiting NF-κB activation and COX-2 expression.
  • Reduces self-renewal and colony-forming capacities of colorectal cancer cells.
  • Ameliorates ulcerative colitis.
  • Significantly inhibits tumor growth in mouse models with low toxicity.
Catalog No. 50-003-88450 Supplier Medchemexpress LLC Supplier No. HYN05895MG
Dehydrodiisoeugenol is an orally active compound demonstrating anti-inflammatory and anti-tumor properties. It is utilized in research focusing on colorectal cancer, inflammatory conditions, and ulcerative colitis.
  • Inhibits proliferation of colorectal cancer cells.
  • Induces apoptosis, autophagy, and cell cycle arrest.
  • Exerts anti-inflammatory effects by inhibiting NF-κB activation and COX-2 expression.
  • Reduces self-renewal and colony-forming capacities of colorectal cancer cells.
  • Ameliorates ulcerative colitis.
  • Significantly inhibits tumor growth in mouse models with low toxicity.