Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dehydroeburicoic acid monoacetate | 77035-42-8 | 5 MG
SDP

Catalog No. 5000418673
Encompass_Preferred

Dehydroeburicoic acid monoacetate is a lanostane triterpenoid isolated from Wolfiporia cocos, intended for research use only. This compound presents as a white to off-white solid with a molecular weight of 510.75 and a chemical formula of C33H50O4.

  • High purity of 97.20%
  • Soluble in DMSO (20 mg/mL) for in vitro applications
  • Can be prepared as a clear solution for in vivo experiments using 10% DMSO and 90% Corn Oil (≥ 2 mg/mL)
  • Recommended storage for solid form: 4°C, sealed, away from moisture and light
  • Recommended storage for solutions: -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Catalog No. 50-004-18673 Supplier Medchemexpress LLC Supplier No. HYN41255MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dehydroeburicoic acid monoacetate is a lanostane triterpenoid isolated from Wolfiporia cocos, intended for research use only. This compound presents as a white to off-white solid with a molecular weight of 510.75 and a chemical formula of C33H50O4.

  • High purity of 97.20%
  • Soluble in DMSO (20 mg/mL) for in vitro applications
  • Can be prepared as a clear solution for in vivo experiments using 10% DMSO and 90% Corn Oil (≥ 2 mg/mL)
  • Recommended storage for solid form: 4°C, sealed, away from moisture and light
  • Recommended storage for solutions: -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.