Learn More
Medchemexpress LLC Dehydroepiandrosterone sulfate (sodium) | 1099-87-2 | >98.0% | 100 MG
SDP
Catalog No. 5000407700
Encompass_Preferred
Dehydroepiandrosterone sulfate (DHEA sulfate; Prasterone sulfate) sodium salt is a neurosteroid and the main secretion product of the adrenal gland. It exhibits non-competitive antagonist activity of GABAA receptor and agonist activity of σ1 receptor. This compound can partially penetrate the blood-brain barrier, making it suitable for neurological research.
  • Inhibits GABAA receptor-mediated chloride influx
  • Enhances NMDA receptor activity through σ1 receptors
  • Exerts anti-inflammatory, anti-glucocorticoid and antidepressant effects
  • Increases convulsive sensitivity
  • Participates in neuroprotection, neurite growth regulation and catecholamine secretion regulation
  • Used in the study of depression, post-traumatic stress disorder (PTSD), and Alzheimer's disease
  • May be a biomarker for cardiovascular disease mortality
Catalog No. 50-004-07700 Supplier Medchemexpress LLC Supplier No. HYB0765100MG
Dehydroepiandrosterone sulfate (DHEA sulfate; Prasterone sulfate) sodium salt is a neurosteroid and the main secretion product of the adrenal gland. It exhibits non-competitive antagonist activity of GABAA receptor and agonist activity of σ1 receptor. This compound can partially penetrate the blood-brain barrier, making it suitable for neurological research.
  • Inhibits GABAA receptor-mediated chloride influx
  • Enhances NMDA receptor activity through σ1 receptors
  • Exerts anti-inflammatory, anti-glucocorticoid and antidepressant effects
  • Increases convulsive sensitivity
  • Participates in neuroprotection, neurite growth regulation and catecholamine secretion regulation
  • Used in the study of depression, post-traumatic stress disorder (PTSD), and Alzheimer's disease
  • May be a biomarker for cardiovascular disease mortality