Learn More
Cayman Chemical Dehydroergosterol, Purity: ≥95%, CAS: 516-85-8, 10 mg
SDP
Catalog No. 502983943
Encompass_Preferred
A fluorescent cholesterol analog (ex/em = 324/375 nm) useful as a probe in membrane research
Catalog No. 50-298-3943 Supplier Cayman Chemical Supplier No. 1970910 MG
A fluorescent cholesterol analog (ex/em = 324/375 nm) useful as a probe in membrane research