Learn More
Medchemexpress LLC Dehydroevodiamine | 67909-49-3 | 5 MG
SDP
Catalog No. 5000421862
Encompass_Preferred
Dehydroevodiamine is a major bioactive quinazoline alkaloid isolated from Evodiae Fructus. It exhibits an antiarrhythmic effect in guinea-pig ventricular myocytes. Additionally, Dehydroevodiamine inhibits LPS-induced iNOS, COX-2, prostaglandin E2 (PGE2), and nuclear factor-kappa B (NF-κB) expression in murine macrophage cells.
  • Isolated from Evodiae Fructus.
  • Exhibits antiarrhythmic effect in guinea-pig ventricular myocytes.
  • Inhibits LPS-induced iNOS and COX-2 expression.
  • Suppresses prostaglandin E2 (PGE2) expression.
  • Inhibits nuclear factor-kappa B (NF-κB) expression in murine macrophage cells.
  • Purity: 99.93%.
  • For research use only.
Catalog No. 50-004-21862 Supplier Medchemexpress LLC Supplier No. HYN21065MG
Dehydroevodiamine is a major bioactive quinazoline alkaloid isolated from Evodiae Fructus. It exhibits an antiarrhythmic effect in guinea-pig ventricular myocytes. Additionally, Dehydroevodiamine inhibits LPS-induced iNOS, COX-2, prostaglandin E2 (PGE2), and nuclear factor-kappa B (NF-κB) expression in murine macrophage cells.
  • Isolated from Evodiae Fructus.
  • Exhibits antiarrhythmic effect in guinea-pig ventricular myocytes.
  • Inhibits LPS-induced iNOS and COX-2 expression.
  • Suppresses prostaglandin E2 (PGE2) expression.
  • Inhibits nuclear factor-kappa B (NF-κB) expression in murine macrophage cells.
  • Purity: 99.93%.
  • For research use only.