Learn More
Medchemexpress LLC Dehydroevodiamine | 67909-49-3 | 99.9% | 301.34 | C19H15N3O | 10 MG
SDP
Catalog No. 5000296398
Encompass_Preferred
Dehydroevodiamine is a quinazoline alkaloid isolated from Evodiae Fructus and supplied for biochemical and pharmacological research. It shows antiarrhythmic activity in guinea-pig ventricular myocytes and reduces LPS-induced iNOS, COX-2, PGE2, and NF-κB expression in macrophage cells.
  • High chemical purity (approximately 99.9%).
  • Molecular formula C19H15N3O; molecular weight 301.34 g/mol.
  • Identified by CAS number 67909-49-3.
  • Available in small research quantities, including 10 mg packs.
  • Suitable for antiarrhythmic and inflammation pathway studies.
Catalog No. 50-002-96398 Supplier Medchemexpress LLC Supplier No. HYN210610MG
Dehydroevodiamine is a quinazoline alkaloid isolated from Evodiae Fructus and supplied for biochemical and pharmacological research. It shows antiarrhythmic activity in guinea-pig ventricular myocytes and reduces LPS-induced iNOS, COX-2, PGE2, and NF-κB expression in macrophage cells.
  • High chemical purity (approximately 99.9%).
  • Molecular formula C19H15N3O; molecular weight 301.34 g/mol.
  • Identified by CAS number 67909-49-3.
  • Available in small research quantities, including 10 mg packs.
  • Suitable for antiarrhythmic and inflammation pathway studies.