Learn More
Cayman Chemical DehydrolIthocholIc AcId 100mg
SDP
Catalog No. 502990572
Encompass_Preferred
A major metabolite of lithocholic acid; formed from LCA by CYP3A4; an agonist of TGR5 (EC50 = 0.27 μM), VDR (EC50 = 3 μM), and FXR in cell-based reporter assays; binds to human PXR (IC50 = 15 μM) and activates mouse and human PXRs in cell-based reporter assays at 100 μM; binds to RORγt (Kd = 1.13 μM for the recombinant human ligand-binding domain) and decreases its activity in a cell-based reporter assay at 10 μM; inhibits the differentiation of TH17 cells at 20 μM
Catalog No. 50-299-0572 Supplier Cayman Chemical Supplier No. 29544100 MG
A major metabolite of lithocholic acid; formed from LCA by CYP3A4; an agonist of TGR5 (EC50 = 0.27 μM), VDR (EC50 = 3 μM), and FXR in cell-based reporter assays; binds to human PXR (IC50 = 15 μM) and activates mouse and human PXRs in cell-based reporter assays at 100 μM; binds to RORγt (Kd = 1.13 μM for the recombinant human ligand-binding domain) and decreases its activity in a cell-based reporter assay at 10 μM; inhibits the differentiation of TH17 cells at 20 μM