Learn More
Cayman Chemical Dehydrolithocholic Acid, Purity: ≥95%, CAS: 1553-56-6, 50 mg
SDP
Catalog No. 502989436
Encompass_Preferred
A major metabolite of lithocholic acid; formed from LCA by CYP3A4; an agonist of TGR5 (EC50 = 0.27 μM), VDR (EC50 = 3 μM), and FXR in cell-based reporter assays; binds to human PXR (IC50 = 15 μM) and activates mouse and human PXRs in cell-based reporter assays at 100 μM; binds to RORγt (Kd = 1.13 μM for the recombinant human ligand-binding domain) and decreases its activity in a cell-based reporter assay at 10 μM; inhibits the differentiation of TH17 cells at 20 μM
Catalog No. 50-298-9436 Supplier Cayman Chemical Supplier No. 2954450 MG
A major metabolite of lithocholic acid; formed from LCA by CYP3A4; an agonist of TGR5 (EC50 = 0.27 μM), VDR (EC50 = 3 μM), and FXR in cell-based reporter assays; binds to human PXR (IC50 = 15 μM) and activates mouse and human PXRs in cell-based reporter assays at 100 μM; binds to RORγt (Kd = 1.13 μM for the recombinant human ligand-binding domain) and decreases its activity in a cell-based reporter assay at 10 μM; inhibits the differentiation of TH17 cells at 20 μM