Learn More
TARGETMOL CHEMICALS INC DEHYDRONITROSONISOLDIPIN 5MG
SDP
Catalog No. 502581605
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dehydronitrosonisoldipine is an inhibitor of sterile (alpha) and TIR motif-containing 1 and can be used in studies about neurodegenerative disorders. Dehydronitrosonisoldipine inhibits axon degenration and the Vincristine-activated cADPR production in neurons. purity: 98%
Catalog No. 50-258-1605 Supplier TARGETMOL CHEMICALS INC Supplier No. T109915MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dehydronitrosonisoldipine is an inhibitor of sterile (alpha) and TIR motif-containing 1 and can be used in studies about neurodegenerative disorders. Dehydronitrosonisoldipine inhibits axon degenration and the Vincristine-activated cADPR production in neurons. purity: 98%