Learn More
Medchemexpress LLC Dehydronuciferine | 7630-74-2 | 5 MG
SDP
Catalog No. 5000419600
Encompass_Preferred
Dehydronuciferine is isolated from the leaves of *Nelumbo nucifera Gaertn* and acts as an acetylcholinesterase (AChE) inhibitor with an IC50 of 25 μg/mL. It also inhibits melanogenesis in mouse B16-4A5 cells.
  • Cholinesterase (ChE) inhibitor
  • Isolated from *Nelumbo nucifera Gaertn*
  • Inhibits melanogenesis in mouse B16-4A5 cells
Catalog No. 50-004-19600 Supplier Medchemexpress LLC Supplier No. HYN42615MG
Dehydronuciferine is isolated from the leaves of *Nelumbo nucifera Gaertn* and acts as an acetylcholinesterase (AChE) inhibitor with an IC50 of 25 μg/mL. It also inhibits melanogenesis in mouse B16-4A5 cells.
  • Cholinesterase (ChE) inhibitor
  • Isolated from *Nelumbo nucifera Gaertn*
  • Inhibits melanogenesis in mouse B16-4A5 cells