Learn More
Medchemexpress LLC Dehydronuciferine | 7630-74-2 | 99.8% | 293.36 g/mol | C19H19NO2 | 10 MG
SDP
Catalog No. 5000289783
Encompass_Preferred
Dehydronuciferine is a natural alkaloid isolated from the leaves of Nelumbo nucifera and characterized as an acetylcholinesterase (AChE) inhibitor. It is provided for research use and has a reported IC50 of 25 μg/mL against AChE.
  • Natural alkaloid isolated from Nelumbo nucifera leaves.
  • Acts as an acetylcholinesterase inhibitor (IC50 25 μg/mL).
  • High purity (≈99.8%).
  • Solid, light yellow to yellow appearance.
  • Storage: 4°C, protect from light; in solvent store at -80°C for long-term storage.
Catalog No. 50-002-89783 Supplier Medchemexpress LLC Supplier No. HYN426110MG
Dehydronuciferine is a natural alkaloid isolated from the leaves of Nelumbo nucifera and characterized as an acetylcholinesterase (AChE) inhibitor. It is provided for research use and has a reported IC50 of 25 μg/mL against AChE.
  • Natural alkaloid isolated from Nelumbo nucifera leaves.
  • Acts as an acetylcholinesterase inhibitor (IC50 25 μg/mL).
  • High purity (≈99.8%).
  • Solid, light yellow to yellow appearance.
  • Storage: 4°C, protect from light; in solvent store at -80°C for long-term storage.