Learn More
Medchemexpress LLC Dehydrothio-p-toluidine | 92-36-4 | MFCD00005780 | 99.9% | 240.32 g/mol | C14H12N2S | 100 G
SDP
Catalog No. 5000208703
Encompass_Preferred
Dehydrothio-p-toluidine (DHPT) is a chemical intermediate used in dyestuff production and related industrial dye synthesis. It is supplied as a light yellow to green yellow solid with a high stated purity and is intended for research or industrial intermediate use; it is not for human or clinical applications.
  • Chemical intermediate for dyestuff production
  • Light yellow to green yellow solid appearance
  • High stated purity (99.9%)
  • Available documentation: datasheet, COA, and SDS
  • Shows inhalation toxicity; handle with appropriate controls
Catalog No. 50-002-08703 Supplier Medchemexpress LLC Supplier No. HYW040269100G
Dehydrothio-p-toluidine (DHPT) is a chemical intermediate used in dyestuff production and related industrial dye synthesis. It is supplied as a light yellow to green yellow solid with a high stated purity and is intended for research or industrial intermediate use; it is not for human or clinical applications.
  • Chemical intermediate for dyestuff production
  • Light yellow to green yellow solid appearance
  • High stated purity (99.9%)
  • Available documentation: datasheet, COA, and SDS
  • Shows inhalation toxicity; handle with appropriate controls