Learn More
Medchemexpress LLC Dehydrothio-p-toluidine | 92-36-4 | MFCD00005780 | 99.9% | 240.33 g/mol | C14H12N2S | 50 G
SDP
Catalog No. 5000209332
Encompass_Preferred
Dehydrothio-p-toluidine is a chemical intermediate used in the production of dyestuffs and related sulfur-containing dyes. Supplied with high reported purity, this material should be handled with care because inhalation toxicity has been reported in acute and subchronic exposures.
  • Used as an intermediate in dyestuff production.
  • Reported purity 99.9% (typical).
  • Available in small pack sizes suitable for laboratory use.
  • CAS number 92-36-4 for unambiguous identification.
  • Handle with appropriate respiratory protection due to inhalation toxicity.
Catalog No. 50-002-09332 Supplier Medchemexpress LLC Supplier No. HYW04026950G
Dehydrothio-p-toluidine is a chemical intermediate used in the production of dyestuffs and related sulfur-containing dyes. Supplied with high reported purity, this material should be handled with care because inhalation toxicity has been reported in acute and subchronic exposures.
  • Used as an intermediate in dyestuff production.
  • Reported purity 99.9% (typical).
  • Available in small pack sizes suitable for laboratory use.
  • CAS number 92-36-4 for unambiguous identification.
  • Handle with appropriate respiratory protection due to inhalation toxicity.