Learn More
Medchemexpress LLC HY-N2490 1mg Medchemexpress, Dehydrotrametenolic acid CAS:6879-05-6 Purity:>98%
SDP
Catalog No. 501970399
Encompass_Preferred
Medchemexpress, HY-N2490 1mg Dehydrotrametenolic acid CAS:6879-05-6 Dehydrotrametenolic acid is a sterol isolated from the sclerotium of Poria cocos. Dehydrotrametenolic acid induces apoptosis through caspase-3 pathway. Dehydrotrametenolic acid has anti-tumor activity, anti-inflammatory, anti-diabetic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0399 Supplier Medchemexpress LLC Supplier No. HYN24901MG
Medchemexpress, HY-N2490 1mg Dehydrotrametenolic acid CAS:6879-05-6 Dehydrotrametenolic acid is a sterol isolated from the sclerotium of Poria cocos. Dehydrotrametenolic acid induces apoptosis through caspase-3 pathway. Dehydrotrametenolic acid has anti-tumor activity, anti-inflammatory, anti-diabetic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.