Learn More
Cayman Chemical DehydrotumulosIc AcId 10mg
SDP
Catalog No. 502910734
Encompass_Preferred
A triterpene with anticancer and anti-inflammatory activities; enhances vincristine-induced cytotoxicity against multidrug-resistant KBv200 cells at 12.5 or 25 UG/ml; reduces serotonin-induced hind paw edema, as well as TPA-induced EAr edema, in mice at 50 mg/kg
Catalog No. 50-291-0734 Supplier Cayman Chemical Supplier No. 3493810 MG
A triterpene with anticancer and anti-inflammatory activities; enhances vincristine-induced cytotoxicity against multidrug-resistant KBv200 cells at 12.5 or 25 UG/ml; reduces serotonin-induced hind paw edema, as well as TPA-induced EAr edema, in mice at 50 mg/kg