Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deionized Formamide, MP Biomedicals™

Catalog No. MP11FORMD00 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP11FORMD00 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP11FORMD00 Supplier MP Biomedicals, Inc Supplier No. 11FORMD002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Deionized Formamide is used in molecular biology and as a denaturing agent for nucleic acids in gel electrophoresis or hybridization experiments.

  • Deionized Formamide is used in molecular biology and a denaturing agent for nucleic acids in gel electrophoresis orr hybridization experiments. In the latter case the role of formamide is to decrease the temperature necessary for the reassociation of complementary nucleic acids.
  • Used in Southern and Northern analyses; 50% or greater concentration for in situ hybridization techniques
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
IUPAC Name formamide
SMILES NC=O

Specifications

Quantity 500 mL
Vapor Pressure 0.08 mm Hg at 20°C
Grade Molecular Biology Grade
Physical Form Solution
Conductivity s100 μS at 25°C
Recommended Storage +4°C, store under nitrogen

Unless specified otherwise, products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.