Learn More
Electron Microscopy Sciences Deionized Water 1 Gal 5 GL
SDP
Catalog No. 5029693
Encompass_Preferred
F.W. 18.01
Reagent Grade Water is typically prepared at 18 megohm/cm specific resistance using a reverse osmosis, mixed deionization, activated filtration and final filtration at 0.2 microns. Color (APHA) <+/-5. Bacteriological purity 0 CFUs/L. Residue after evaporation 10ppm. Coliform negative.
Catalog No. 50-296-93 Supplier Electron Microscopy Sciences Supplier No. 2280005
F.W. 18.01
Reagent Grade Water is typically prepared at 18 megohm/cm specific resistance using a reverse osmosis, mixed deionization, activated filtration and final filtration at 0.2 microns. Color (APHA) <+/-5. Bacteriological purity 0 CFUs/L. Residue after evaporation 10ppm. Coliform negative.