Learn More
Description
- Resistivity greather than 15 MΩ at 25°C
Specifications
Specifications
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 18 g/mol |
| Percent Purity | High-purity |
| Grade | Reagent |
| Packaging | Plastic Bottle |
| Quantity | 1 gal. |
| Chemical Name or Material | Water |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.