Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Water, Deionized, ASTM Type III (ASTM D1193) (10L)
SDP

Catalog No. NC0467154
Encompass_Preferred

Water, Deionized, ASTM Type III (ASTM D1193) (10L)

Catalog No. NC0467154 Supplier Aqua Solutions Supplier No. 976410L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water, Deionized, ASTM Type III (ASTM D1193) (10L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.