Learn More
Inorganic Ventures Deionized Water Blank
SDP
Catalog No. 507527058
Encompass_Preferred
Inorganic Ventures' Deionized Water Blank is a 500mL reference material set in a water matrix for stability. This ICP blank is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-752-7058 Supplier Inorganic Ventures Supplier No. IVDIBLANK500ML
Inorganic Ventures' Deionized Water Blank is a 500mL reference material set in a water matrix for stability. This ICP blank is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search