Learn More
Thermo Scientific™ Deionizer Cartridges
Catalog No. 0903492
Change view
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Porous Strong Ion Exchange
Strong Anion Exchange
Strong Cation Exchange
Two Bed Ion Exchange
Ultrapure, Oxygen Removal
Ultrapure, Oxygen and Organic Removal
Capacity:
1680 grains
1760 grains
2000 gal.
30 g
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
5000 gal.
5000 gal. (as CaCO3)
785 grains/1000 gal.
875 grains
For Use With (Application):
Anion Removal
Cation Removal
High capacity, two-bed ion exchange
Organic Removal
Oxygen Removal
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure, Oxygen and Organic Removal
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Resin Type Capacity For Use With (Application)
09-034-92 Bantam™ Deionizer, B-Pure™ Strong Anion Exchange 1680 grains Anion Removal
11-164-58 B-Pure™ Activated Carbon 5000 gal. (as CaCO3) Organic Removal
11-164-57 B-Pure™ Mixed Bed - Ultrapure
09-034-3 Bantam™ Deionizer, B-Pure™ Mixed Bed 875 grains -
09-034-8 B-Pure™ Porous Strong Ion Exchange 30 g Oxygen Removal
09-034-5 Bantam™ Deionizer, B-Pure™ Activated Carbon 5000 gal. Organic Removal
09-034-10 Bantam™ Deionizer, B-Pure™ Strong Cation Exchange 3170 grains Cation Removal
09-028-33 Bantam™ Deionizer, B-Pure™ Mixed Bed, Activated Carbon 785 grains/1000 gal. Ultrapure and Organic Removal
09-034-104 B-Pure™ Macroreticular, Activated Carbon 5000 gal. Pretreatment
09-034-5C B-Pure™ Macroreticular, Activated Carbon 2000 gal. Pretreatment Macropure
09-050-47 B-Pure™ Extruded Carbon 5000 gal. Organic Removal
09-050-52 B-Pure™ Ultrapure, Oxygen Removal 455 grains/30 g Ultrapure and Organic Removal
09-034-90 B-Pure™ Ultrapure, Oxygen and Organic Removal 365 grains/2000 gal./12 g Ultrapure, Oxygen and Organic Removal
09-034 Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated Two Bed Ion Exchange 1760 grains High capacity, two-bed ion exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 09-034-92 Supplier Thermo Scientific™ Supplier No. D0760
Add to Cart
Add to Cart
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
TRUSTED_SUSTAINABILITY
Specifications
Description Cartridge for anion removal; strong anion exchange; full size
Capacity 1680 grains
Resin Type Strong Anion Exchange
For Use With (Application) Anion Removal
For Use With (Equipment) Bantam™ Deionizer, B-Pure™
Type Deionizer Cartridge