Learn More
Sklar Instruments Dejerine Hammer, Sklar 8 in., Blunt, Simple, Hollow Grip, Stainless Steel / Rubber, EA.
SDP
Catalog No. 501181220 Shop All Sklar Instruments Products
Encompass_Preferred
Dejerine Hammer, Sklar. 8 in., Blunt, Simple, Hollow Grip, Stainless Steel / Rubber. Sklar's Dejerine Simple Percussion Hammer is a diagnostic instrument used to test reflexes. The head of the instrument features equal size, dual bumpers. It is cylindrical, non-latex material encircled by metal in the middle which ultimately forms the neck of the instrument. The neck becomes wider and forms the handle. The handle is commonly hollow and grooved for a better grip. The overall length is 8 inches.
Catalog No. 50-118-1220 Supplier Sklar Instruments Supplier No. 063250
Dejerine Hammer, Sklar. 8 in., Blunt, Simple, Hollow Grip, Stainless Steel / Rubber. Sklar's Dejerine Simple Percussion Hammer is a diagnostic instrument used to test reflexes. The head of the instrument features equal size, dual bumpers. It is cylindrical, non-latex material encircled by metal in the middle which ultimately forms the neck of the instrument. The neck becomes wider and forms the handle. The handle is commonly hollow and grooved for a better grip. The overall length is 8 inches.