Learn More
Aviva Systems Biology Corp DEK Antibody - N-terminal region (P100637_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100637_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DEK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DEK. Alternate names: D6S231EApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-62673