Learn More
Medchemexpress LLC HY-18932 5mg Medchemexpress, DEL-22379 CAS:181223-80-3 Purity:>98%
SDP
Catalog No. 502082092
Encompass_Preferred
Medchemexpress, HY-18932 5mg DEL-22379 CAS:181223-80-3 DEL-22379 is an ERK dimerization Inhibitor. DEL-22379 readily binds to ERK2 with a Kd estimated in the low micromolar range, though binding is detectable even at low nanomolar concentrations. ERK2 dimerization is progressively inhibited with an IC50 of ~0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2092 Supplier Medchemexpress LLC Supplier No. HY189325MG
Medchemexpress, HY-18932 5mg DEL-22379 CAS:181223-80-3 DEL-22379 is an ERK dimerization Inhibitor. DEL-22379 readily binds to ERK2 with a Kd estimated in the low micromolar range, though binding is detectable even at low nanomolar concentrations. ERK2 dimerization is progressively inhibited with an IC50 of ~0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.