Learn More
Roboz Surgical DEL OP SCS 4.75IN STR S/B
SDP
Catalog No. NC9199218
Encompass
Delicate Operating Scissors; Straight; Sharp-Blunt; 30mm Blade Length; 4-3/4 in. Overall Length
Catalog No. NC9199218 Supplier Roboz Surgical Supplier No. RS6700
Delicate Operating Scissors; Straight; Sharp-Blunt; 30mm Blade Length; 4-3/4 in. Overall Length