Learn More
Roboz Surgical DEL OP SCS 4.75IN STR S/S
SDP
Catalog No. NC9344278
Encompass
Delicate Operating Scissors; Straight; Sharp-Sharp; 30mm Blade Length; 4-3/4 in. Overall Length
Catalog No. NC9344278 Supplier Roboz Surgical Supplier No. RS6702
Delicate Operating Scissors; Straight; Sharp-Sharp; 30mm Blade Length; 4-3/4 in. Overall Length