Learn More
Cayman Chemical DelafloxacIn 10mg
SDP
Catalog No. 502984411
Encompass_Preferred
A fluoroquinolone antibiotic that demonstrates extremely high potency against a wide range of bacteria in vitro; has an anionic structure, which enhances its potency in acidic environments and in skin and soft tissue infections
Catalog No. 50-298-4411 Supplier Cayman Chemical Supplier No. 2031410 MG
A fluoroquinolone antibiotic that demonstrates extremely high potency against a wide range of bacteria in vitro; has an anionic structure, which enhances its potency in acidic environments and in skin and soft tissue infections